5-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
H54497.MDEA 85.3 EUR
ALFAH54497.MD ALFAH54497.03
5-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040203