N,N'-Diallyl-L-tartardiamide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI EUROPE
Warning

D1539-25G
TCIAD1539-25GEA 66 EUR
TCIAD1539-25G
N,N'-Diallyl-L-tartardiamide ≥98.0% (by HPLC, titration analysis)
N,N'-Diallyl-L-tartardiamide
Formula: C₁₀H₁₆N₂O₄
MW: 228,25 g/mol
Número MDL: MFCD00008640
Núm. CAS: 58477-85-3
EINECS: 261-277-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c761e0d18a5663496905eceb7c44c56e9f21d79&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Bromination Method) min. 98.0 %
Specific rotation [a]20/D +105 to +110 deg (C=2.4, H2O)
Melting point 180.0 to 185.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR