Cholesterol hexyl carbonate
Proveedor: TCI EUROPE
TCIAC0725-10GEA
276
EUR
TCIAC0725-10G
Cholesterol hexyl carbonate
Cholesterol hexyl carbonate
Formula:
C₃₄H₅₈O₃ |
Número MDL:
MFCD00059504 Núm. CAS: 15455-80-8 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Specific rotation [a]20/D | -29.0 to -33.0 deg (C=2,CHCl3) |
Solubility in Chloroform | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...