Cholesterol lauryl carbonate ≥95.0% (by HPLC)
Proveedor: TCI EUROPE
TCIAC1025-1GEA
112
EUR
TCIAC1025-1G
Cholesterol lauryl carbonate ≥95.0% (by HPLC)
Cholesterol lauryl carbonate
Formula:
C₄₀H₇₀O₃ |
Número MDL:
MFCD00059910 Núm. CAS: 15455-85-3 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Mesomorphic range | 53.0 to 80.0 deg-C |
Specific rotation [a]20/D | -23.5 to -27.5 deg (C=2, CHCl3) |
Solubility in Chloroform | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...