Flame-retardant rain jacket, Baltero 9461
Proveedor: Sioen
Sio-Start® FR
113-7320EA 249 EUR
113-7320 113-7318 113-7317 113-7319 113-7316 113-7315
Flame-retardant rain jacket, Baltero 9461
Ropa de trabajo
This waterproof and windproof jacket is made of Sio-Start FR AST material, consisting of 100% polyester fabric with FR PU coating + AST. It features a fixed flame-retardant cotton lining and an additional detachable flame-retardant fleece lining. The outer fabric is water-repellent, while the coating on the inside attracts moisture and keeps the body warm and dry.
  • Highly breathable, supple, high tear resistance
  • Straight collar with foldaway hood
  • Zip closure under flap with touch & close fastening
  • Two patched pockets, one Napoleon pocket
  • Sleeve narrowing by press studs
Back length: 85 cm
Weight: 240 g/m²
Certificaciones: EN 13034 (Type PB/6), EN 531 (A/B2/C1), EN 14116, EN 1149-5, EN 343 (3-3)