High visibility T-shirts, Gardhar 2666
Proveedor: Sioen
113-7028EA 39.4 EUR
SIOE2666AMCFR1S SIOE2666AMCFR3XL SIOE2666AMCFR1M SIOE2666AMCFR1L SIOE2666AMCFR1XL SIOE2666AMCFR2XL
High visibility T-shirts, Gardhar 2666
This lightweight shirt made of 100% Sio-Cool® polyester piqué features a round neck and short sleeves. It is machine washable up to 40 °C.
  • Solid, supple, with stitched seams
  • Welded, segmented reflective tape
  • Wicks perspiration away
  • Nano sized silver particles: Permanent anti-microbial featurs eliminate all odours
Weight: 180 g/m²
Certificaciones: EN 471 (2-2), EN 13758-2 + A1 (UPF 50+)