Drawer organiser trays, Drawer/Ganizers®
Proveedor: Spectrum
SPEC155525EA 84 EUR
SPEC155525 SPEC155645 SPEC155495
Drawer organiser trays, Drawer/Ganizers®
Polystyrene.
  • Durable
  • Protect laboratory supplies
  • Promote laboratory safety
  • Provides secondary containment
Drawer/Ganizers portable organizer trays is designed for transporting assorted lab items and mobilizing experimental activities.