Evaporating dishes
Proveedor: Avignon
216-2730EA 177 EUR
AVIG10111 AVIG10113B AVIG10106B AVIG10109 AVIG10106 AVIG10117 AVIG10105 AVIG10107 AVIG10102 AVIG10101 AVIG10115 AVIG10114 AVIG10103
Evaporating dishes
Evaporating dishes, porcelain, with spout.
  • Excellent temperature resistance
  • With minimal thermal expansion
  • High chemical resistance
  • High resistance to thermal shocks