Goggles, B-Line
Proveedor: Bollé Safety
B-Line
111-0982EA 4.6 EUR
111-0982 111-0981
Goggles, B-Line
Gafas /Googles
B-line is a safety eyewear range designed to protect wearers from industrial risks.
  • PVC frame
  • With or without anti-fog coating
  • Elastic strap
Certificaciones: EN 166 / EN 170