Accessories for LLE columns, CHROMABOND® XTR
Proveedor: MACHEREY-NAGEL
MANA730508EA 885 EUR
MANA730508
Accessories for LLE columns, CHROMABOND® XTR
Columnas de cromatografía
A variable polypropylene rack is recommended for the liquid-liquid extraction (LLE) with CHROMABOND® XTR columns.
  • For application of up to 24 columns
  • Flow by gravity (no vacuum)