4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAC1713-1GEA
533
EUR
TCIAC1713-1G
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC, titration analysis)
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula:
C₁₅H₂₀O₇ MW: 312,3151 g/mol |
Número MDL:
MFCD00143276 Núm. CAS: 56683-55-7 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...