Ácido DIPSO ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
TCIAB1494-25GEA
38
EUR
TCIAB1494-25G
Ácido DIPSO ≥98.0% (by HPLC, titration analysis)
DIPSO
Formula:
C₇H₁₇NO₆S MW: 261,29 g/mol |
Número MDL:
MFCD00038353 Núm. CAS: 68399-80-4 EINECS: 269-992-2 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Absorbance | max. 0.15 (0.5mol/L、H2O、260nm) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...