Alcohol 3,5-di-terc-butil-4-hidroxibencilico ≥97.0% (por GC)

Proveedor: TCI EUROPE
Atención

TCIAD1480-25GEA 199 EUR
TCIAD1480-25G
Alcohol 3,5-di-terc-butil-4-hidroxibencilico ≥97.0% (por GC)
Alcohol 3,5-di-terc-butil-4-hidroxibencilico
Formula: C₁₅H₂₄O₂
MW: 236,35 g/mol
Número MDL: MFCD00017254
Núm. CAS: 88-26-6
EINECS: 201-815-6
https://es.vwr.com/stibo/web/std.lang.all/83/06/28978306.jpg

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 139.0 to 142.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR