Ácido (4-formilfenoxi)acético ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAF0319-25GEA
149
EUR
TCIAF0319-25G
Ácido (4-formilfenoxi)acético ≥97.0% (by HPLC, titration analysis)
Ácido (4-formilfenoxi)acético
Formula:
(C₆H₄(CHO))OCH₂COOH MW: 180,16 g/mol Pto. de fusión: 215 °C |
Número MDL:
MFCD00016613 Núm. CAS: 22042-71-3 EINECS: 244-749-3 |
Resultados test especificaciones
Appearance | White to Light red to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in hot Methanol | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...