Metilurea ≥98.0% (by total nitrogen basis)

Proveedor: TCI EUROPE

Sinónimos: Metilcarbamida

TCIAM0455-500GEA 55 EUR
TCIAM0455-500G TCIAM0455-25G
Metilurea ≥98.0% (by total nitrogen basis)
Metilurea
Formula: C₂H₆N₂O
MW: 74,08 g/mol
Pto. de fusión: 98…102 °C
Número MDL: MFCD00007950
Núm. CAS: 598-50-5
EINECS: 209-935-0
https://es.vwr.com/stibo/web/std.lang.all/38/99/28973899.jpg

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 96.0 to 102.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR