4-Methoxyphthalic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAM1432-1GEA
24
EUR
TCIAM1432-1G
TCIAM1432-5G
4-Methoxyphthalic acid ≥98.0% (by GC, titration analysis)
4-Methoxyphthalic acid
Formula:
C₉H₈O₅ |
Número MDL:
MFCD00094739 Núm. CAS: 1885-13-8 EINECS: 217-546-2 |
![]() |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...