4'-Chloro-2'-methylacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI EUROPE
TCIAC2104-25GEA
51
EUR
TCIAC2104-25G
4'-Chloro-2'-methylacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Chloro-2'-methylacetoacetanilide
Formula:
C₁₁H₁₂ClNO₂ |
Número MDL:
MFCD00018493 Núm. CAS: 20139-55-3 EINECS: 243-540-4 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...