Methyl (4-fluorophenyl)acetate 99%
Proveedor: Thermo Scientific
|
Atención
|
ACRO304670250EA
152
EUR
ACRO304670250
Methyl (4-fluorophenyl)acetate 99%
Methyl (4-fluorophenyl)acetate
Formula:
C₉H₉FO₂ MW: 168,17 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00800608 Núm. CAS: 34837-84-8 |
Resultados test especificaciones
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4860 to 1.4880 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...