3',4',5-Trichlorosalicylanilide ≥98.0% (by titrimetric analysis)

Proveedor: TCI EUROPE

TCIAT0668-10GEA 333 EUR
TCIAT0668-10G
3',4',5-Trichlorosalicylanilide ≥98.0% (by titrimetric analysis)
3',4',5-Trichlorosalicylanilide
Formula: C₁₃H₈Cl₃NO₂
Número MDL: MFCD00059622
Núm. CAS: 642-84-2
EINECS: 211-391-4
https://es.vwr.com/stibo/web/std.lang.all/48/37/28994837.jpg

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR