o-Anisidinium chloride ≥99.0% (by HPLC)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAA0489-25GEA
68
EUR
TCIAA0489-25G
o-Anisidinium chloride ≥99.0% (by HPLC)
o-Anisidinium chloride
Formula:
C₇H₁₀ClNO MW: 159,6134 g/mol |
Número MDL:
MFCD00054334 Núm. CAS: 134-29-2 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...