2-Fluoro-4-(methylthio)benzeneboronic acid, 98%
H53064.MDEA 172 EUR
ALFAH53064.MD
2-Fluoro-4-(methylthio)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 957060-84-3
Molecular Formula: C7H8BFO2S
Formula Weight: 186.01
MDL No.: MFCD09027247