N-Benzyl-4-methoxyaniline ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
TCIAB0409-5GEA
69
EUR
TCIAB0409-5G
N-Benzyl-4-methoxyaniline ≥98.0% (by GC, titration analysis)
N-Benzyl-4-methoxyaniline
Formula:
C₁₄H₁₅NO MW: 213,28 g/mol Pto. Ebullición: 238°C |
Número MDL:
MFCD00059298 Núm. CAS: 17377-95-6 ADR: 6.1,III |
![]() |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 46.0 to 50.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...