3,5-Dichloro-N-(2-fluorobenzyl)aniline, 97%

H57048.MDEA 61 EUR
ALFAH57048.MD ALFAH57048.03
3,5-Dichloro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 723753-74-0
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD05740291
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR