2-Fluoro-N-(3-fluorobenzyl)-4-methylaniline, 97%

H57738.03EA 94.7 EUR
ALFAH57738.03 ALFAH57738.MD
2-Fluoro-N-(3-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1040049-97-5
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR