Tribromsalan ≥97.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAT0356-25GEA
46
EUR
TCIAT0356-25G
Tribromsalan ≥97.0% (by titrimetric analysis)
Tribromsalan
Formula:
C₁₃H₈Br₃NO₂ |
Número MDL:
MFCD00045740 Núm. CAS: 87-10-5 EINECS: 201-723-6 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 224.0 to 228.0 deg-C |
Solubility in hot Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...