4'-Chloro-2',5'-dimethoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI EUROPE

TCIAC2100-25GEA 29 EUR
TCIAC2100-25G
4'-Chloro-2',5'-dimethoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Chloro-2',5'-dimethoxyacetoacetanilide
Formula: C₁₂H₁₄ClNO₄
Número MDL: MFCD00026256
Núm. CAS: 4433-79-8
EINECS: 224-638-6
https://es.vwr.com/stibo/web/std.lang.all/91/55/28939155.jpg

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR