Dry box gloves, PU
Proveedor: Piercan
112-5280EA 418 EUR
112-5280
Dry box gloves, PU
Guantes
These PU gloves provide protection against mechanical risks of tear strength, perforation and breaking. They are ideal for applications in high oxidising environment including ozone, active oxygen and free radicals.
  • Exceptional mechanical properties (puncture, tear, cut)
  • High resistance to ozone and UV
  • Flexibility and dexterity
  • Excellent behavior against ionising radiations
Suitable fields of application include nuclear industries and life sciences (pharmaceutical/medical).
Certificaciones: EN 388:2016 (3X22X), EN 421; Cat. III